
5-Hexyn-1-ol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:928-90-5
:
Formula:C₆H₁₀O
MDL Number:MFCD00002980
:98.14 g/mol
Boiling Pt:73…75 °C (15 mmHg)
Density:0.880
Flash Pt:70 °C (158 °F)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together