4-Hydroxycinnamic acid
Glass bottle for solids, 100 g
Catalog # MOLE86815064-100G
Marketsource
Supplier: Molekula
4-Hydroxycinnamic acid
Catalog # MOLE86815064-100G
Supplier: Molekula
Marketsource
CAS Number: 7400-08-0
Jump to:
Product Details & Documents
Synonyms: p-Coumaric acid, p-Cumaric acid, 3-(4-Hydroxyphenyl)acrylic acid, p-Hydroxy-Cinnamic Acid
- CAS Number:7400-08-0
- Formula:C₉H₈O₃
- MDL Number:MFCD00004399
- Molecular Weight:164.16 g/mol
- EINECS:231-000-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



