
3-Methoxycinnamic acid
Catalog # MOLE57097526-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:178.19 g/mol
EINECS:228-049-5
:GHS07
CAS Number:6099-04-3
Formula:C₁₀H₁₀O₃
MDL Number:MFCD00004386