

5-Hexen-1-ol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:821-41-0
:
Formula:C₆H₁₂O
MDL Number:MFCD00002981
:100.16 g/mol
Boiling Pt:152…155 °C
Melting Pt:< –20 °C
Density:0.842
Flash Pt:59 °C (138 °F)
Storage Temperature:Ambient
EINECS:212-477-4
UN:2924
ADR:3,III
:GHS05
:GHS02
Related Information
Frequently Bought Together