
5-Hexen-1-ol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:821-41-0
:
Formula:C₆H₁₂O
MDL Number:MFCD00002981
:100.16 g/mol
Boiling Pt:152…155 °C (760 mmHg)
Melting Pt:-20 °C
Density:0,8725 g/cm³
Flash Pt:47 °C
Storage Temperature:Raumtemperatur
EINECS:212-477-4
UN:1987
ADR:3,III
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4340 to 1.4360 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together