0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrochemistry Meters
Oxygen Meters
VWR®, Accessories for DO Meter, DO 40
VWR®, Accessories for DO Meter, DO 40
VWR®, Accessories for DO Meter, DO 40
Catalog # 664-0235
Supplier:
VWR Collection
Expand image
VWR®, Accessories for DO Meter, DO 40
Catalog # 664-0235
Supplier:
VWR Collection
Specifications
Description:
Maintenance kit for DO 40, incl. 3 spare membranes and 100 ml electrolyte
Pk:
1
Specifications
About this item
Product Details
Product Details