Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Oleic acid

Marketsource
Supplier:   Roth Carl
Oleic acid
Supplier:  Roth Carl
Marketsource
Certificates
Oleic acid

2 Options Available Below

Product Details & Documents
Synonyms:  (9Z)-9-Octadecenoic acid
  • CAS Number:112-80-1
  • Formula:C₁₈H₃₄O₂
  • MDL Number:MFCD00064242
  • Molecular Weight:282.47 g/mol
  • Merck Index:13,06898
  • Boiling Pt:286 °C (1013 hPa)
  • Melting Pt:16.7 °C
  • Density:0.8946 g/cm³ (20 °C)
  • Flash Pt:180 °C
  • Storage Temperature:Freezer
  • EINECS:204-007-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 mL
    Log in for availability
    Log in for availability
    1 * 5 mL
    1 * 5 mL - SFr. 47.40
    SFr. 47.40
    1 * 5 mL
       SFr. 47.40

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    50 mL
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - SFr. 342.00
    SFr. 342.00
    1 * 50 mL
       SFr. 342.00

    Specifications:
    Marketsource
    Marketsource