Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Oleic acid

Glass bottle, 50 mL
Catalog # ROTH6812.1
Marketsource
Supplier:   Roth Carl
Oleic acid
Catalog # ROTH6812.1
Supplier:  Roth Carl
Marketsource
CAS Number:  112-80-1
Oleic acid
Oleic acid

Glass bottle, 50 mL

CAS Number:  112-80-1
Price
Inquire For Price
Log in for availability
1 * 50 mL
Product Details & Documents
Synonyms:  (9Z)-9-Octadecenoic acid
  • CAS Number:112-80-1
  • Formula:C₁₈H₃₄O₂
  • MDL Number:MFCD00064242
  • Molecular Weight:282.47 g/mol
  • Merck Index:13,06898
  • Boiling Pt:286 °C (1013 hPa)
  • Melting Pt:16.7 °C
  • Density:0.8946 g/cm³ (20 °C)
  • Flash Pt:180 °C
  • Storage Temperature:Freezer
  • EINECS:204-007-1
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL

Specifications