Acrylamide ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98+% CAS No.: 79-06-1 Molecular Formula: CH2=CHCONH2 Formula Weight: 71.08 Flash Point: 138°(280°F) Odour: Odourless Boiling Point: 125°/25mm Beilstein Reference: 605349 Merck Reference: 14,129 UN No.: 2074 MDL No.: MFCD00008032
Synonyms: Prop-2-enamide
- CAS Number:79-06-1
- Formula:C₃H₅NO
- MDL Number:MFCD00008032
- Molecular Weight:71.08 g/mol
- Merck Index:14,129
- Boiling Pt:125 °C (25 mmHg)
- Melting Pt:82…85 °C
- Density:1.322
- Flash Pt:138 °C (280 °F)
- Storage Temperature:Ambient
- EINECS:201-173-7
- UN:2074
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 79-06-1 Molecular Formula: CH2=CHCONH2 Formula Weight: 71.08 Flash Point: 138°(280°F) Odour: Odourless Boiling Point: 125°/25mm Beilstein Reference: 605349 Merck Reference: 14,129 UN No.: 2074 MDL No.: MFCD00008032
Synonyms: Prop-2-enamide
- CAS Number:79-06-1
- Formula:C₃H₅NO
- MDL Number:MFCD00008032
- Molecular Weight:71.08 g/mol
- Merck Index:14,129
- Boiling Pt:125 °C (25 mmHg)
- Melting Pt:82…85 °C
- Density:1.322
- Flash Pt:138 °C (280 °F)
- Storage Temperature:Ambient
- EINECS:201-173-7
- UN:2074
- ADR:6.1,III
Documents
Organic Building Blocks and Intermediates
Application Notes
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



