Order Entry
Switzerland
ContactUsLinkComponent
Tetrachloroethylene ≥99.9%, SPECTRONORM® for spectroscopy
Health
Environment
Tetrachloroethylene ≥99.9%, SPECTRONORM® for spectroscopy
Supplier:  VWR Chemicals
Certificates
undefined
Health
Environment
Tetrachloroethylene ≥99.9%, SPECTRONORM® for spectroscopy
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (GC):
    ≥ 99.9 %
  • Acidity:
    ≤ 0.0005 meq/g
  • Alkalinity:
    ≤ 0.0005 meq/g
  • Colouration:
    ≤ 10 APHA
  • Evaporation residue:
    ≤ 10 ppm
  • Water:
    ≤ 100 ppm
  • Absorbance (290 nm):
    ≤ 0.70
  • Absorbance (295 nm):
    ≤ 0.19
  • Absorbance (300 nm):
    ≤ 0.10
  • Absorbance (305 nm):
    ≤ 0.20
  • Absorbance (350 nm):
    ≤ 0.05
  • Absorbance (400 nm):
    ≤ 0.03
  • Transmittance (290 nm):
    ≥ 20 %
  • Transmittance (295 nm):
    ≥ 65 %
  • Transmittance (300 nm):
    ≥ 80 %
  • Transmittance (305 nm):
    ≥ 85 %
  • Transmittance (350 nm):
    ≥ 89 %
  • Transmittance (400 nm):
    ≥ 95 %
  • Fluorescence (as quinine) (365 nm):
    ≤ 1.0 ppb

Specification Test Results

Related Information

Frequently Bought Together