
3-Cyclohexylpropionic acid 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:701-97-3
:
Formula:C₉H₁₆O₂
MDL Number:MFCD00001527
:156.22 g/mol
Boiling Pt:275.7 °C (1013 hPa)
Melting Pt:14…17 °C
Density:0.998 g/cm³
Flash Pt:113 °C
EINECS:211-861-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4630 to 1.4650 (20°C, 589 nm)
- Titration with NaOH:≥98.5 %
Specification Test Results
Frequently Bought Together