



2,2,4-Trimethylpentane, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
:540-84-1
:
:(CH₃)₂CHCH₂C(CH₃)₃
:MFCD00008943
:114.23 g/mol
:13,05212
:99.2 °C (1013 hPa)
:–107 °C
:0.692 g/cm³ (20 °C)
:Ambient
:208-759-1
:1262
:3,II
:M-Clarity™ quality level = MQ300
:–12 °C
:GHS02
:GHS08
:GHS07
:GHS09
- Solubility:0.56 mg/l (25 °C)
- Melting Point:-107 °C
- Molar Mass:114.23 g/mol
- Boiling Point:99 °C (1013 hPa)
- Vapor Pressure:51 hPa (20 °C)
- Flash Point:-12 °C
- Refractive Index:2.3
- Explosion Limit:1 %(V)
- Density:0.69 g/cm3 (20 °C)
- pH:(H2O) neutral
- Ignition Point:410 °C
- Saturation concentration:239 g/m3 (20 °C)
Frequently Bought Together