

Isoamyl alcohol ≥98.5% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123-51-3
:
Formula:(CH₃)₂CHCH₂CH₂OH
MDL Number:MFCD00002934
:88.15 g/mol
Merck Index:14,5195
Boiling Pt:130…132 °C
Melting Pt:–117 °C
Density:0.809
Flash Pt:45 °C (113 °F)
Storage Temperature:Ambient
EINECS:204-633-5
UN:1105
ADR:3,II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Related Information
Frequently Bought Together