


Isobutanol
Supplier: Roth Carl
Certificates
About this item
CAS Number:78-83-1
:
Formula:(CH₃)₂CHCH₂OH
MDL Number:MFCD00004740
:74.12 g/mol
Merck Index:13,05148
Boiling Pt:107.9 °C (1013 hPa)
Melting Pt:–108 °C
Density:0.802 g/cm³ (20 °C)
Flash Pt:28 °C
Storage Temperature:Ambient
EINECS:201-148-0
UN:1212
ADR:3,III
:GHS02
:GHS05
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Frequently Bought Together