


Isobutanol ≥98%, GPR RECTAPUR®
Catalog # 20836.297
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay:Min. 98 %
- Boiling point:107 - 109 °C
- Density (20/4):0.799 - 0.803
- Evaporation residue:Max. 0.02 %
- Refractive index (20°C):1.395 - 1.396
Specification Test Results
About this item
:
:74.12 g/mol
Merck Index:13,05148
Boiling Pt:107.9 °C (1013 hPa)
Melting Pt:–108 °C
Density:0.802 g/cm³ (20 °C)
Flash Pt:28 °C
EINECS:201-148-0
UN:1212
ADR:3,III
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:78-83-1
Formula:(CH₃)₂CHCH₂OH
MDL Number:MFCD00004740