


Isobutanol, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
1.00984.2500
: Merck
:
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Solubility:85 g/l (20 °C)
- Melting Point:-108 °C
- Molar Mass:74.12 g/mol
- Boiling Point:106 - 108 °C (1013 hPa)
- Vapor Pressure:12 hPa (20 °C)
- Flash Point:28 °C
- Explosion Limit:1.5 - 12 %(V)
- Density:0.802 g/cm3 (20 °C)
- pH:7 (80 g/l, H2O, 20 °C)
- Ignition Point:430 °C
- Evaporation number:24
:
:74.12 g/mol
:13,05148
:107.9 °C (1013 hPa)
:–108 °C
:0.802 g/cm³ (20 °C)
:28 °C
:Ambient
:201-148-0
:1212
:3,III
:M-Clarity™ quality level = MQ300
:GHS02
:GHS05
:GHS07
:78-83-1
:(CH₃)₂CHCH₂OH
:MFCD00004740