Order Entry
Switzerland
ContactUsLinkComponent
tert-Butyl methyl ether for synthesis, Supelco®
Flammable
Irritant
tert-Butyl methyl ether for synthesis, Supelco®
Catalog # 8.18109.9025
Supplier:  Merck
CAS Number:  
undefined
Flammable
Irritant
tert-Butyl methyl ether for synthesis, Supelco®
Catalog # 8.18109.9025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    25 L
  • Seq:
    5327

Specifications

About this item

 :
 :88.15 g/mol
Merck Index:13,06059
Boiling Pt:55.2 °C (1013 hPa)
Melting Pt:–108.6 °C
Density:0.7405 g/cm³ (20 °C)
Flash Pt:–28 °C
Storage Temperature:Ambient
EINECS:216-653-1
UN:2398
ADR:3,II
 :GHS02
 :GHS07
CAS Number:1634-04-4
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812