Order Entry
Switzerland
ContactUsLinkComponent
AMP (2-Amino-2-methylpropanol) 99%
Irritant
AMP (2-Amino-2-methylpropanol) 99%
Certificates
undefined
Irritant
AMP (2-Amino-2-methylpropanol) 99%
Certificates

Specification Test Results

  • Appearance:
    Colorless Low melting solid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.8 %
  • Titration with HClO4:
    ≥98.5 %
  • UV:
    10 cm cell vs water
  • Color scale:
    ≤10 APHA
  • Suitability:
    (in liquid scint. counting) Passes test

Specification Test Results

Related Information

Frequently Bought Together