2-Propylvaleric acid for synthesis, Sigma-Aldrich®
Catalog # 8.14439.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 ml
- Seq:2315
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:2 g/l (20 °C)
- Molar Mass:144.21 g/mol
- Boiling Point:220 °C (1013 hPa)
- Vapor Pressure:0.01 hPa (20 °C)
- Flash Point:111 °C
- Explosion Limit:1 %(V)
- Density:0.905 g/cm3 (20 °C)
Specification Test Results
About this item
Synonyms: 2-Propylpentanoic acid, Valproic Acid, 2,2-Di-n-propylacetic acid
:
:144.21 g/mol
Merck Index:12,10049
Boiling Pt:220 °C (1013 hPa)
Density:0.905 g/cm³ (20 °C)
Flash Pt:111 °C
Storage Temperature:Ambient
EINECS:202-777-3
UN:3265
ADR:8,III
:M-Clarity™ quality level = MQ200
:GHS08
:GHS07
CAS Number:99-66-1
Formula:C₈H₁₆O₂
MDL Number:MFCD00002672