Order Entry
Switzerland
ContactUsLinkComponent
2-Ethylcaproic acid for synthesis, Sigma-Aldrich®
Health
2-Ethylcaproic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Health
2-Ethylcaproic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    1.4 g/l (20 °C)
  • Melting Point:
    -59 °C
  • Molar Mass:
    144.21 g/mol
  • Boiling Point:
    226 - 229 °C (1013 hPa)
  • Vapor Pressure:
    0.04 hPa (20 °C)
  • Flash Point:
    114 °C
  • Refractive Index:
    1.4241 (20 °C, 589 nm)
  • Explosion Limit:
    0.9 - 6.7 %(V)
  • Density:
    0.91 g/cm3 (20 °C)
  • pH:
    3 (1.4 g/l, H2O, 20 °C)
  • Ignition Point:
    310 °C

Specification Test Results

Related Information

Frequently Bought Together