ContactUsLinkComponent
J.T.Baker® BAKERBOND Speedisk® Quanternary Amine, SPE Columns
J.T.Baker® BAKERBOND Speedisk® Quanternary Amine, SPE Columns
Catalog # 8168-02
Supplier:  Avantor
J.T.Baker® BAKERBOND Speedisk® Quanternary Amine, SPE Columns
Catalog # 8168-02
Supplier:  Avantor
Specifications
  • Capacity:
    1 ml
  • Bed weight:
    35 mg
  • Phase:
    Q-amine
  • Pk:
    1
Specifications
About this item
Ultra-clean, PP columns with BAKERBOND® HPLC media are engineered to save time, minimise work and final solvent volumes and improve performance. Run times are up to nine times faster than conventional columns. The built-in column guard filter and higher analyte concentration may reduce or eliminate pre-filtration and concentration steps.
  • Run times up to nine times faster than conventional SPE columns
  • Unique laminar configuration
  • Silica based
General applications: Ionic, acidic analytes.
These SPE columns can be used in strong ion exchange mode.