



n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:66…69 °C(1.013 hPa)
Melting Pt:–95 °C
Density:0.660…0.668 g/cm³ (20 °C)
Flash Pt:7.4 %(V)
EINECS:203-777-6
UN:1208
ADR:3,II
:GHS07
:GHS08
:GHS02
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Vapor Density:~3 (vs air)
- Assay (GC):Min. 97 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.01 %
- Free acid (as CH₃COOH):Max. 0.001 %
- Free alkali (as NH₃):Max. 0.0005 %
- Calcium (Ca):Max. 0.00001 %
- Potassium (K):Max. 0.00001 %
- Magnesium (Mg):Max. 0.00001 %
- Sodium (Na):Max. 0.00001 %
- Transmittance at 200 nm:Min. 20 %
- Transmittance at 225 nm:Min. 80 %
- Transmittance from 250 nm:Min. 98 %
- Suitability for HPLC-MS:complying
- Refractive Index:n20/D 1.375 (lit.)
- Vapor Pressure:~132 mmHg ( 20 °C)
Specification Test Results
Frequently Bought Together