Order Entry
Switzerland
ContactUsLinkComponent
1-Butanesulphonic acid sodium salt for synthesis, Sigma-Aldrich®
1-Butanesulphonic acid sodium salt for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
1-Butanesulphonic acid sodium salt for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Melting Point:
    310 °C
  • Molar Mass:
    160.17 g/mol

Specification Test Results

Related Information

Frequently Bought Together