
1-Butanesulphonic acid sodium salt 99+% for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2386-54-1
:
Formula:C₄H₉NaO₃S
MDL Number:MFCD00007540
:160.17 g/mol
Melting Pt:>300 °C
EINECS:219-201-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 % (1 g, 105°C, 2 hrs)
- Solubility:(10 % in water) Clear to slightly hazy colorless
- UV:(5.5 % in water, pH 3) 1 cm cell
- Titration after Ion exchange:≥99.0 % (On dry substance)
Specification Test Results
Frequently Bought Together