2-Mercaptoethanol
Glass bottle for solids, 250 g
Catalog # MOLE37807035-250G
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 2-Hydroxyethyl mercaptan, β-Mercaptoethanol
- CAS Number:60-24-2
- Formula:HSCH₂CH₂OH
- MDL Number:MFCD00004890
- Molecular Weight:78.14 g/mol
- Merck Index:13,05894
- Boiling Pt:157 °C (1013 hPa)
- Melting Pt:–100 °C
- Density:1.12 g/cm³ (20 °C)
- Flash Pt:68 °C
- Storage Temperature:Ambient
- EINECS:200-464-6
- UN:2966
- ADR:6.1,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



