Order Entry
Switzerland
ContactUsLinkComponent
Sodium methoxide for synthesis, Sigma-Aldrich®
Flammable
Corrosive
Sodium methoxide for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Corrosive
Sodium methoxide for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Melting Point:
    127 °C (decomposition)
  • Molar Mass:
    54.02 g/mol
  • Bulk Density:
    500 - 600 kg/m3
  • Boiling Point:
    300 °C
  • Vapor Pressure:
    (20 °C)
  • Density:
    1.3 g/cm3 (20 °C)
  • Ignition Point:
    240 °C
  • Synonyms:
    Sodium methoxide, Sodium methanolate
  • Hill Formula:
    CH3NaO
  • Chemical Formula:
    CH3ONa

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together