Order Entry
Switzerland
ContactUsLinkComponent
Methanol absolute acetone-free
Methanol absolute acetone-free
Catalog # M1775-1GA
   Safer Human and Environmental Health
CAS Number:  
undefined
Methanol absolute acetone-free
Catalog # M1775-1GA
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1
  • Seq:
    8080
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion
  • Pk Info:
    1 gal

Specifications

Specification Test Results

  • Vapor Pressure:
    97.68 mmHg ( 20 °C)
  • Appearance:
    complying
  • Autoignition Temperature:
    725 °F
  • Refractive Index:
    n20/D 1.329 (lit.)
  • Explosion Limit:
    36 %

Specification Test Results

About this item

 :
 :32.04 g/mol
Merck Index:13,05984
Boiling Pt:64…65 °C (1.013 hPa)
Melting Pt:–98 °C
Density:0.79 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
 :GHS08
 :GHS02
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595