Order Entry
Switzerland
ContactUsLinkComponent
 
Methanol ≥99.6%, Laboratory Reagent
    
 
undefined
Methanol ≥99.6%, Laboratory Reagent
    

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

 

 

  • Explosion Limit:
    36 %
  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.6 %
  • Non-volatile matter:
    Max. 0.01 %
  • Water (Karl Fischer):
    Max. 0.1 %
  • Free acid (as HCOOH):
    Max. 0.03 %
  • Free alkali (as NH₃):
    Max. 0.01 %
  • Vapor Pressure:
    410 mmHg ( 50 °C)
  • Autoignition Temperature:
    725 °F
  • Vapor Density:
    1.11 (vs air)

 

 

Frequently Bought Together