Order Entry
Switzerland
ContactUsLinkComponent
Methanol ≥99.9%, PESTINORM® for capillary GC analysis
Flammable
Toxic
Health
Methanol ≥99.9%, PESTINORM® for capillary GC analysis
Catalog # 83966.320
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  
undefined
Flammable
Toxic
Health
Methanol ≥99.9%, PESTINORM® for capillary GC analysis
Catalog # 83966.320
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.9 %
  • Acidity:
    Max. 0.0005 meq/g
  • Water:
    Max. 0.1 %
  • Evaporation residue (100°C):
    Max. 0.0005 %
  • Organic residue (as Octanol) (GC/FID):
    Max. 10 ng/l
  • Halogenated residue (as Lindane)(GC/ECD):
    Max. 5 ng/l

Specification Test Results

About this item

 :
 :32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3 (6.1),II
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
REACH:01-2119433307-44
 :GHS02
 :GHS06
 :GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595