Order Entry
Switzerland
ContactUsLinkComponent
Methanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Flammable
Toxic
Health
Methanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Flammable
Toxic
Health
Methanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.9 %
  • Carbonyl compounds:
    Passes test
  • Acidity:
    Max. 0.0002 Meq/g
  • Alkalinity:
    Max. 0.0002 Meq/g
  • Density (20/20):
    0.791 --> 0.793
  • Evaporation residue:
    Max. 3 ppm
  • Water:
    Max. 0.03 %
  • Ca (Calcium):
    Max. 200 ppb
  • K (Potassium):
    Max. 100 ppb
  • Mg (Magnesium):
    Max. 100 ppb
  • Na (Sodium):
    Max. 200 ppb
  • Gradient (235 nm):
    Max. 2 MAU
  • Gradient (254 nm):
    Max. 2 MAU
  • Transmittance (205 nm):
    Min. 10 %
  • Transmittance (210 nm):
    Min. 40 %
  • Transmittance (220 nm):
    Min. 60 %
  • Transmittance (230 nm):
    Min. 80 %
  • Transmittance (240 nm):
    Min. 90 %
  • Transmittance (from 260 nm):
    Min. 98 %
  • Fluorescence (as quinine) (254 nm):
    Max. 1 ppb
  • Fluorescence (as quinine) (365 nm):
    Max. 1 ppb
  • Conforms to ACS:
    Passes test
  • Filtered through 0.2 μm:
    Confirmed

Specification Test Results

Frequently Bought Together