Order Entry
Switzerland
ContactUsLinkComponent
Ethanol (untaxed) ≥96% (v/v), Puriss., meets analytical specification of BP, Ph. Eur., Riedel-de Haën™
Flammable
Irritant
Ethanol (untaxed) ≥96% (v/v), Puriss., meets analytical specification of BP, Ph. Eur., Riedel-de Haën™
Certificates
Flammable
Irritant
Ethanol (untaxed) ≥96% (v/v), Puriss., meets analytical specification of BP, Ph. Eur., Riedel-de Haën™
Certificates

Specification Test Results

  • Appearance:
    complying
  • Identity A:
    complying
  • Identity B:
    complying
  • Boiling range:
    78 - 79 °C
  • Vapor Density:
    1.59 (vs air)
  • Density (D 20/20):
    0.805 - 0.812
  • Water (Karl Fischer):
    4.8 - 7.4 %
  • Non-volatile matter:
    Max. 0.001 %
  • Acidity or alkalinity:
    complying
  • Methanol (GC):
    Max. 200 Vol.ppm
  • Volatile impurities (GC):
    complying
  • Residual solvents:
    complying
  • UV-absorption:
    complying
  • Explosion Limit:
    19 %, 60 °F
  • Refractive Index:
    n20/D 1.3600 (lit.)

Specification Test Results

Related Information

Frequently Bought Together