Order Entry
Switzerland
ContactUsLinkComponent
2-Propanol ≥99.5% ACS, Riedel-de Haën™
Flammable
Irritant
2-Propanol ≥99.5% ACS, Riedel-de Haën™
Catalog # 673773-4L
   Safer Human and Environmental Health
CAS Number:  
undefined
Flammable
Irritant
2-Propanol ≥99.5% ACS, Riedel-de Haën™
Catalog # 673773-4L
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    4 L
  • Seq:
    5162
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Vapor Pressure:
    44 mmHg ( 25 °C)
  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.50 %
  • Solubility in water:
    complying
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.2 %
  • Free acid, free alkali:
    Max. 0.0001 meq/g
  • Acetone (GC):
    Max. 0.002 %
  • Carbonyl compounds (as propionaldehyde):
    Max. 0.002 %
  • APHA:
    Max. 10
  • Refractive Index:
    n20/D 1.377 (lit.)

Specification Test Results

About this item

 :
 :60.1 g/mol
Merck Index:13,05228
Boiling Pt:77…83 °C (1.013 hPa)
Melting Pt:–90 °C
Density:0.784…0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
REACH:01-2119457558-25
 :GHS02
 :GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674