1-Hexanol
Marketsource
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Hexyl alcohol, Pentyl carbinol, 1-Hexyl alcohol, 1-Hydroxy hexane
- CAS Number:111-27-3
- Formula:H₃C(CH₂)₄CH₂OH
- MDL Number:MFCD00002982
- Molecular Weight:102.18 g/mol
- Merck Index:13,04715
- Boiling Pt:156 °C (1013 hPa)
- Melting Pt:–45 °C
- Density:0.8322 g/cm³ (20 °C)
- Flash Pt:60 °C
- Storage Temperature:Ambient
- EINECS:203-852-3
- UN:2282
- ADR:3,III
2 Options Available Below
Product Details & Documents
Synonyms: Hexyl alcohol, Pentyl carbinol, 1-Hexyl alcohol, 1-Hydroxy hexane
- CAS Number:111-27-3
- Formula:H₃C(CH₂)₄CH₂OH
- MDL Number:MFCD00002982
- Molecular Weight:102.18 g/mol
- Merck Index:13,04715
- Boiling Pt:156 °C (1013 hPa)
- Melting Pt:–45 °C
- Density:0.8322 g/cm³ (20 °C)
- Flash Pt:60 °C
- Storage Temperature:Ambient
- EINECS:203-852-3
- UN:2282
- ADR:3,III



