

Bis(2-methoxyethyl)ether stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:111-96-6
:
Formula:C₆H₁₄O₃
MDL Number:MFCD00008503
Merck Index:13,03187
Boiling Pt:162 °C (1013 hPa)
Melting Pt:–64 °C
Density:0.944 g/cm³ (20 °C)
Flash Pt:53 °C
EINECS:203-924-4
UN:3271
ADR:3,III
:M-Clarity™ quality level = MQ200
:134.18 g/mol
:GHS02
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-64 °C
- Molar Mass:134.17 g/mol
- Boiling Point:155 - 165 °C (1013 hPa)
- Vapor Pressure:2 hPa (20 °C)
- Flash Point:70 °C
- Refractive Index:1.4097 (20 °C, 589 nm)
- Explosion Limit:1.4 - 17.4 %(V)
- Density:0.944 g/cm3 (20 °C)
- pH:(H2O, 20 °C) neutral
- Ignition Point:190 °C
- Evaporation number:90
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
Frequently Bought Together