Order Entry
Switzerland
ContactUsLinkComponent
Ethylene glycol monomethyl ether (2-methoxyethanol)
Flammable
Health
Irritant
Ethylene glycol monomethyl ether (2-methoxyethanol)
Catalog # EHERC13328200
Supplier:  EHRENSTORFER
CAS Number:  
Flammable
Health
Irritant
Ethylene glycol monomethyl ether (2-methoxyethanol)
Catalog # EHERC13328200
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :76.1 g/mol
Merck Index:12,06118
Boiling Pt:124.6 °C (1013 hPa)
Melting Pt:–85 °C
Density:0.964 g/cm³ (20 °C)
Flash Pt:39 °C
EINECS:203-713-7
UN:1188
ADR:3,III
 :GHS02
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:109-86-4
Formula:C₃H₈O₂
MDL Number:MFCD00002867