


Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:109-86-4
:
Formula:C₃H₈O₂
MDL Number:MFCD00002867
:76.1 g/mol
Merck Index:14,6038
Boiling Pt:124…125 °C
Melting Pt:–85 °C
Density:0.965
Flash Pt:46 °C (115 °F)
Storage Temperature:Ambient
EINECS:203-713-7
UN:1188
ADR:3,III
:GHS08
:GHS02
:GHS07
Related Information
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
Thermo Scientific Organic Photovoltaics
Product Literature
Frequently Bought Together