Order Entry
Switzerland
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, 2-Ethoxyethanol
Catalog # 79109-5ML-F
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, 2-Ethoxyethanol
Catalog # 79109-5ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    110-80-5
  • Description:
    2-Ethoxyethanol
  • Seq:
    02
  • Environmentally Preferable:
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H226 - H302 - H331 - H360FD
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00002869
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Ethyl glycol; Ethylene glycol monoethyl ether
  • Melting point:
    –90 °C (lit.)
  • Formula:
    C₂H₅OCH₂CH₂OH
  • Molecular weight:
    90,12
  • Density:
    0,93 g/ml at 25 °C (lit.)
  • Flash point:
    40 °C
  • Boiling point:
    135 °C (lit.)

Specifications

About this item

  • GHS signal: GHS02, GHS06, GHS08
Caution: P202 - P210 - P233 - P301 + P312 - P304 + P340 + P311 - P308 + P313