


N-Ethyldiisopropylamine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7087-68-5
:
Formula:C₈H₁₉N
MDL Number:MFCD00008868
:129.25 g/mol
Boiling Pt:126…127 °C
Melting Pt:–46 °C
Density:0.747
Flash Pt:8 °C (46 °F)
Storage Temperature:Ambient
EINECS:230-392-0
UN:3384
ADR:6.1,I
:GHS02
:GHS06
:GHS05
Related Information
Frequently Bought Together