Order Entry
Switzerland
ContactUsLinkComponent
Dipropylamine for synthesis, Sigma-Aldrich®
Flammable
Toxic
Corrosive
Dipropylamine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Toxic
Corrosive
Dipropylamine for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    35 g/l (25 °C) (experimental)
  • Melting Point:
    -63 °C
  • Molar Mass:
    101.19 g/mol
  • Boiling Point:
    105 °C (1013 hPa)
  • Vapor Pressure:
    38 hPa (20 °C)
  • Flash Point:
    7 °C
  • Refractive Index:
    1.4049 (20 °C, 589 nm)
  • Explosion Limit:
    1.8 - 9.3 %(V)
  • Density:
    0.74 g/cm3 (20 °C)
  • pH:
    (1 g/l, H2O, 25 °C)
  • Ignition Point:
    260 °C

Specification Test Results

Related Information

Frequently Bought Together