Ethylenediamine
Catalog # WAKO169-21443
Supplier: Wako
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:60.1 g/mol
Merck Index:13,03829
Boiling Pt:117 °C (1013 hPa)
Melting Pt:8.5 °C
Density:0.898 g/cm³ (20 °C)
Flash Pt:34 °C
Storage Temperature:Ambient
EINECS:203-468-6
UN:1604
ADR:8,II
:GHS02
:GHS05
:GHS08
:GHS07
CAS Number:107-15-3
Formula:C₂H₈N₂
MDL Number:MFCD00008204