Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), Supelco®
SUPL93440-50G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
- Environmentally Preferable:
: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM
:
:121.14 g/mol
:357
:201-064-4
:GHS07
:169…172 °C
:Ambient
:77-86-1
:NH₂C(CH₂OH)₃
:MFCD00004679
:1.328 g/cm³ (20 °C)
:13,09842