Order Entry
Switzerland
ContactUsLinkComponent
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Catalog # MOLEM56787308
Supplier:  Molekula
CAS Number:  
undefined
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Catalog # MOLEM56787308
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM
 :
 :121.14 g/mol
Boiling Pt:357
EINECS:201-064-4
 :GHS07
Melting Pt:169…172 °C
Storage Temperature:Ambient
CAS Number:77-86-1
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679
Density:1.328 g/cm³ (20 °C)
Merck Index:13,09842