Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), Millipore®
: Merck
: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM
:77-86-1
:
:NH₂C(CH₂OH)₃
:MFCD00004679
:121.14 g/mol
:13,09842
:357 °C (1013 hPa)
:169…172 °C
:1.328 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:Ambient
:201-064-4
- Environmentally Preferable:
Frequently Bought Together