Order Entry
Switzerland
ContactUsLinkComponent
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99%, Electran for electrophoresis
Irritant
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99%, Electran for electrophoresis
Catalog # 443866G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99%, Electran for electrophoresis
Catalog # 443866G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • Assay:
    Min. 99.00%
  • Appearance:
    White or colorless crystal powder
  • Identity (IR):
    Passes test
  • DNases:
    Not detected
  • RNases:
    Not detected
  • Proteases:
    Not detected
  • Absorbance (260 nm) (10%):
    Max. 0.030
  • Absorbance (280 nm) (10%):
    Max. 0.020
  • pH (20 °C; 1 mol/l):
    10.50 - 11.50
  • Heavy metals (as Pb):
    Max. 0.005%
  • Water:
    Max. 0.3%
  • Cu (Copper):
    Max. 0.0001%
  • Fe (Iron):
    Max. 0.0001%
  • Pb (Lead):
    Max. 0.0001%

Specification Test Results

About this item

 :
 :121.14 g/mol
Merck Index:13,09842
Boiling Pt:357 °C (1013 hPa)
Melting Pt:169…172 °C
Density:1.328 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-064-4
 :GHS07
CAS Number:77-86-1
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679