Order Entry
Switzerland
ContactUsLinkComponent
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8-100.5%, AnalaR NORMAPUR® analytical reagent
Irritant
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8-100.5%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8-100.5%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (calculated on anhydrous):
    99.8 - 100.5 %
  • Appearance:
    White crystalline powder
  • Identification:
    Passes test
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Ignition residue (SO4):
    Max. 100 ppm
  • Insolubility in water:
    Max. 30 ppm
  • Water:
    Max. 0.1 %
  • Cl (Chloride):
    Max. 5 ppm
  • SO4 (Sulphate):
    Max. 5 ppm
  • As (Arsenic):
    Max. 0.2 ppm
  • Ca (Calcium):
    Max. 10 ppm
  • Cd (Cadmium):
    Max. 0.1 ppm
  • Cu (Copper):
    Max. 0.1 ppm
  • Fe (Iron):
    Max.1 ppm
  • K (Potassium):
    Max. 1 ppm
  • Mg (Magnesium):
    Max. 0.3 ppm
  • Na (Sodium):
    Max. 5 ppm
  • Pb (Lead):
    Max. 0.1 ppm
  • Absorbance (280 nm) (0.5 mol/l):
    Max. 0.05

Specification Test Results

Frequently Bought Together