Order Entry
Switzerland
ContactUsLinkComponent
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.9%, Ultrapure
Irritant
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.9%, Ultrapure
Supplier:  PanReac AppliChem
Certificates
undefined
Irritant
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.9%, Ultrapure
Supplier:  PanReac AppliChem

Specification Test Results

  • Assay (titr.):
    min. 99.9 %
  • pH (1 M; H2O; 20°C):
    10.5 - 11.5
  • Water:
    max. 0.2 %
  • Cu:
    max. 0.0001 %
  • Fe:
    max. 0.0001 %
  • PB:
    max. 0.0001 %
  • A (1 cm/1 M in H2O) at 430 nm:
    max. 0.004
  • A (1 cm/1 M in H2O) at 260 nm:
    0.03
  • A (1 cm/1 M in H2O) at 280 nm:
    0.02

Specification Test Results

Frequently Bought Together