

3-Aminopropan-1-ol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:156-87-6
:
Formula:C₃H₉NO
MDL Number:MFCD00008223
:75.11 g/mol
Boiling Pt:184…187 °C
Melting Pt:11 °C
Density:0.983
Flash Pt:101 °C (214 °F)
Storage Temperature:Ambient
EINECS:205-864-4
UN:2735
ADR:8,II
:GHS05
:GHS07
Related Information
Frequently Bought Together