Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Ethanolamine ≥98%

Ethanolamine ≥98%

Corrosive
Irritant
Ethanolamine ≥98%
Certificates
Ethanolamine ≥98%
Ethanolamine ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 141-43-5 Molecular Formula: C2H7NO Formula Weight: 61.08 Flash Point: 93°(199°F) Refractive Index: 1.4540 Odour: Ammonia-like Melting Point: 10° Boiling Point: 169-170° Beilstein Reference: 505944 Merck Reference: 14,3727 UN No.: 2491 MDL No.: MFCD00008183

Synonyms:  2-Aminoethanol, Monoethanolamine
  • CAS Number:141-43-5
  • Formula:NH₂CH₂CH₂OH
  • MDL Number:MFCD00008183
  • Molecular Weight:61.08 g/mol
  • Merck Index:14,3727
  • Boiling Pt:169…170 °C
  • Melting Pt:10 °C
  • Density:1.012
  • Flash Pt:93 °C (199 °F)
  • Storage Temperature:Ambient
  • EINECS:205-483-3
  • UN:2491
  • ADR:8,III
Product Family Options

Product Information