ContactUsLinkComponent
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) 99+%
Irritant
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) 99+%
Certificates
undefined
Irritant
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) 99+%
Specification Test Results
  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤1.0 % (K.F.)
  • Titration with HClO4:
    ≥99.0 %