Order Entry
Switzerland
ContactUsLinkComponent
Triethanolamine (trolamine) 99.0-103.0% (by acidimetry), SAFC®
Triethanolamine (trolamine) 99.0-103.0% (by acidimetry), SAFC®
Certificates
undefined
Triethanolamine (trolamine) 99.0-103.0% (by acidimetry), SAFC®
Certificates

Specification Test Results

  • Assay (acidimetric, calc. on anhydrous substance):
    99.0 - 103.0 %
  • Identity (IR-spectrum):
    passes test
  • Identity (GC):
    conforms
  • Appearance:
    colorless to slightly yellow viscose liquid
  • Appearance of solution:
    Clear and not more intense in colour than reference solution B₆
  • Clarity of solution:
    passes test
  • PH (1.0 g in 10 ml water):
    10.5 - 11.5
  • Refractive index (n 20/D):
    1.482 - 1.485
  • Relative Density (d 20/20):
    1.120 - 1.130
  • Relative Density (d 25/25):
    1.120 - 1.128
  • Heavy metals (as Pb):
    ≤ 0.0010 %
  • As (Arsenic):
    ≤ 0.000075 %
  • Fe (Iron):
    ≤ 0.0005 %
  • Related substances (Ethanolamine, GC):
    ≤ 0.1 %
  • Related substances (Diethanolamine, GC):
    ≤ 0.5 %
  • Related substances (total impurity, GC):
    ≤ 1.0 %
  • Related substances (N-Nitrosodiethanolamine, GC):
    ≤ 24 ppb
  • Residual solvents (ICH Q3C):
    ...excluded by production process
  • Sulfated ash (600 °C):
    ≤ 0.05 %
  • Water (according to Karl Fischer):
    ≤ 0.5 %
  • Endotoxins:
    ≤ 2.5 I.U./ml

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together